Would you like to log in again?
Recombinant Human IL-16 (121 a.a.) (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7151-U002 21-7151-U500 21-7151-U100 21-7151-U010
10821-156 10821-162 10821-160 10821-158
Recombinant Human IL-16 (121 a.a.) (Carrier-free), Tonbo Biosciences
IL-16 is an immunomodulatory cytokine secreted by activated CD8+ T cells, as well as other immune and non-immune cell types. It serves as a chemoattractant for CD4+ T cells, macrophages, and eosinophils. IL-16 induces expression of CD25 (IL-2R) and plays a role in the accumulation and activation of CD4+ cells recruited to sites of inflammation. IL-16 (121 a.a.) is a proteolytic product of pro-IL-16 cleaved by Caspase-3 after T-cell activation, producing a secreted biologically active form. It requires surface CD4 to mediate signaling processes.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty