Would you like to log in again?
Recombinant Human IL-13 Variant (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7150-U002 21-7150-U010 21-7150-U100 21-7150-U500
10821-148 10821-150 10821-152 10821-154
Recombinant Human IL-13 Variant (Carrier-free), Tonbo Biosciences
IL-13, produced primarily by Th2 cells and to a lesser extent by mast and NK cells, is an immunoregulatory cytokine. Its various functions include induction of matrix metalloproteinases, enhancement of IgE secretion from activated B cells and B cell proliferation. Additionally, IL-13 plays a role in the inhibition of the expression of inflammatory cytokines and modulates resistance to various parasites. IL-13 is also secreted by eosinophils and in the lung is essential in regulating eosinophilic inflammation, mucus secretion, and airway hyperresponsiveness. Its function as a central mediator of allergen-induced asthma is well documented. Single nucleotide polymorphisms result in variants associated with specific asthmatic or allergic phenotypes, and certain variants will show enhanced functional activity as compared to wild-type IL-13.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty