Human Recombinant Gitrl (from E. coli)
Supplier: Tonbo Biosciences
21-7146-U005 21-7146-U100 21-7146-U500 21-7146-U020
10821-120EA 910.25 USD
10821-120 10821-122 10821-116 10821-118
Human Recombinant Gitrl (from E. coli)
GITRL, also known as TNFSF18 and AITRL, along with its corresponding receptor, are members of the TNF superfamily. GITRL is expressed in endothelial cells and is thought to modulate T lymphocyte survival in peripheral tissues. It plays a role in interactions between activated T lymphocytes and endothelial cells. GITRL also triggers an increase in the capability of endothelial cells to support leukocyte adhesion.