Would you like to log in again?
Recombinant Human Chemerin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7145-U005 21-7145-U500 21-7145-U100 21-7145-U025
10821-108EA 86.98 USD InStock
10821-108 10821-114 10821-112 10821-110
Recombinant Human Chemerin (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Chemerin, an adipokine and member of the Cystatin superfamily, is highly expressed in white adipose tissue. It is a ligand for the GPCR ChemR23 (CMKLR1, DEZ) and it acts as a chemoattractant for cells expressing this receptor. Chemerin is a secreted protein that, after cleavage by serine protease, becomes an active protein that can be found in plasma and serum. Studies show bioactive chemerin generation occurs during the early stages of inflammation, implicating a role as a bridge between innate and adaptive immunity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty