Would you like to log in again?
Recombinant Human BAFF Receptor (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7136-U050 21-7136-U500 21-7136-U100 21-7136-U010
10821-038 10821-042 10821-040 10821-036
Recombinant Human BAFF Receptor (Carrier-free), Tonbo Biosciences
The B-cell activating factor (BAFF) receptor is a type III transmembrane protein belonging to the TNFR superfamily. It is expressed in spleen, lymph node and resting B-cells, with limited expression in activated B cells, resting CD4+ cells and peripheral blood leukocytes. BAFF/BAFFR activities are crucial to the survival and maturation of peripheral B cells. BAFF binding to BAFFR expressed on a subset of T cells has been shown to play a role in costimulating T cell activation.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty