Would you like to log in again?
Recombinant Human 4-1BB Ligand (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7135-U020 21-7135-U005 21-7135-U500 21-7135-U100
10821-030EA 218.26 USD InStock
10821-030 10821-028 10821-034 10821-032
Recombinant Human 4-1BB Ligand (Carrier-free), Tonbo Biosciences
Proteins and Peptides
4-1BB Ligand, also known as TNFSF9, is a type II surface glycoprotein expressed on activated T-cells and antigen presenting cells (APCs). As a member of the TNF superfamily, it functions in promoting CD4+ and CD8+ T cell cytokine production. 4-1BB/4-1BBL signaling also enhances cell proliferation and T cell effector function. A recent study shows that 4-1BBL expressed on T cells can restrict effector T cell development under tolerogenic conditions.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty