Would you like to log in again?
Recombinant Human Serpin F1 (PEDF) (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7131-U100 21-7131-U020 21-7131-U500 21-7131-U005
10820-936EA 778.5 USD InStock
10820-936 10820-934 10820-938 10820-932
Recombinant Human Serpin F1 (PEDF) (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Serpin F1, also known as PEDF, is a secreted glycoprotein that belongs to the clade F subfamily of the serpin family of proteinase inhibitors. Because it does not display any serine protease inhibitory activity it belongs to a second group, separate from other serpin family members that do possess typical protease inhibitor function. Serpin F1 is expressed in many tissues including lung, liver, skin, heart and ovary, and it has diverse functions. These include anti-angiogenic, neuroprotective, and anti-carcinogenic activities. The protein has been mapped and different regions have been associated with its differing roles. It binds to the PEDF receptor, and may have other receptor targets.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty