Would you like to log in again?
Recombinant Human Relaxin-3 (INSL7) (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7129-U100 21-7129-U025 21-7129-U005 21-7129-U500
10820-928 10820-926 10820-924 10820-930
Recombinant Human Relaxin-3 (INSL7) (Carrier-free), Tonbo Biosciences
At least seven relaxin-like proteins make up the Insulin/Relaxin superfamily. Most consist of two peptide subunits, an alpha-chain and a beta-chain, linked by disulfide bonds. Relaxin-3, also known as INSL7, has been reported as a ligand for LGR7 (RXFP1), and is the only known ligand for GPCR135 (RXFP3). It binds less often with GPCR142. Relaxin-3 is a secreted protein that is reported to play a role in stress response. It is almost exclusively expressed within the brain stem.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty