Would you like to log in again?
Recombinant Human R-Spondin 1 (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7126-U500 21-7126-U020 21-7126-U100 21-7126-U005
10820-906EA 1994.99 USD InStock
10820-906 10820-902 10820-904 10820-900
Recombinant Human R-Spondin 1 (Carrier-free), Tonbo Biosciences
Proteins and Peptides
R-Spondin 1 (RSPO1) is a secreted protein belonging to the R-Spondin family of four structurally related Wnt/beta-catenin signaling regulators. All four members contain one thrombospondin-like and two furin-like domains. During early development, R-Spondin 1 is thought to play a role in neural tube development. It has also been shown to be involved in female gonad determination. R-Spondins can enhance Wnt/beta-catenin signaling as they compete with the Wnt agonist DKK-1 for binding to Wnt co-receptors LRP6/Kremen. They can also act as a signaling activation ligand on LRP6 and LGR4 to form a cluster with WNT and FZD. The R-Spondins have been demonstrated to bind to LGR-4, LGR-5 and LGR-6 receptors with high affinity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty