Would you like to log in again?
Recombinant Mouse Prolactin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7125-U100 21-7125-U050 21-7125-U010 21-7125-U500
10820-896EA 315.9 USD InStock
10820-896 10820-894 10820-892 10820-898
Recombinant Mouse Prolactin (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Prolactin is a globular, secreted neuroendocrine pituitary hormone present as both glycosylated and non-glycosylated monomers. The non-glycoslyated form is expressed in the pituitary gland and stored in granules prior to being secreted. The glycosylated protein reportedly has lower biologic activity but is constitutively secreted. Prolactin binds the prolactin receptor (PRLR), which has widespread expression. Prolactin is a lactogenic protein, acting mainly on the mammary gland and promoting lactation in females during pregnancy and suckling. It also has been reported to have functions related to immunoregulation, breast and prostate cancer development, and regulation of reproductive function.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty