Would you like to log in again?
Recombinant Human Pleiotrophin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7123-U020 21-7123-U500 21-7123-U100 21-7123-U005
10820-878 10820-882 10820-880 10820-876
Recombinant Human Pleiotrophin (Carrier-free), Tonbo Biosciences
The novel heparin-binding cytokine pleiotrophin (also known as HB-GAM) is a neurotrophic factor, structurally related to midkine. This protein is generally found in the central and peripheral nervous systems, and expression is highly regulated during development. Neurogenesis and cell migration are some of the proposed functions of pleiotrophin. Like midkine, it signals through the anaplastic lymphoma kinase (ALK) receptor leading to MAPK pathway activation. Pleiotrophin also associates with N-syndecan, enhancing axonal growth and neurite extension.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty