Would you like to log in again?
Recombinant Human Persephin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7122-U020 21-7122-U500 21-7122-U005 21-7122-U100
10820-870 10820-874 10820-868 10820-872
Recombinant Human Persephin (Carrier-free), Tonbo Biosciences
Persephin (PSP) is a secreted protein belonging to the family of neurotrophic factors that includes GDNF, neurturin (NTN) and artemin, all distantly related to the TGF-beta superfamily. Neurotrophic factors play in a role in development and maintenance of the nervous system. Persephin is reported to promote both the survival and growth of central dopaminergic and motor neurons, and kidney development, but does not support the peripheral nervous system. It is expressed at very low levels in many tissues. Persephin does not seem to signal through the same receptor complex as other family members GDNF and neurturin, but uses subunit GFR alpha-4 along with the receptor tyrosine kinase RET.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty