Would you like to log in again?
Recombinant Mouse PDGF-BB (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7120-U100 21-7120-U010 21-7120-U002
10820-858 10820-856 10820-854
Recombinant Mouse PDGF-BB (Carrier-free), Tonbo Biosciences
Platelet-derived growth factor-BB (PDGF-BB) is one of the five dimeric proteins belonging to the PDGF family. Other members are homodimers PDGF-AA, PDGF-CC, PDGF-DD, and the heterodimer PDGF-AB. The PDGFs are produced by platelets, stored in platelet alpha-granules and are released upon platelet activation. PDGF proteins are generally considered as potent mitogens for connective tissue cells, and can also be chemotactic for fibroblasts, smooth muscle cells, neutrophils and mononuclear cells. PDGF-BB plays a role in angiogenesis and smooth muscle formation during embryonic development, as well as wound healing and regeneration in adults. Two receptors have been identified for the PDGF proteins - PDGFR alpha and PDGFR beta. PDGF-BB binds to PDGFR alpha, as well as PDGFR beta which appears to be specific for PDGF-BB and PDGF-AB.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty