Human Recombinant Neurturin (from E. coli)
Supplier: Tonbo Biosciences
21-7118-U005 21-7118-U500 21-7118-U020 21-7118-U100
10820-840EA 221.7 USD
10820-840 10820-844 10820-842 10820-838
Human Recombinant Neurturin (from E. coli)
Neurturin (NTN) is a secreted protein belonging to the family of neurotrophic factors that includes GDNF, persephin (PSP) and artemin, all distantly related to the TGF-beta superfamily. Similar to GDNF, neurturin is a secreted protein that exhibits strong trophic activity on dopaminergic neurons. It also promotes the development and survival of sympathetic and sensory neurons. Neurturin signals through the receptor tyrosine kinase (RET)/GFR-alpha 2 receptor complex, although it can also bind GFR-alpha 1 at higher concentrations.