Would you like to log in again?
Recombinant Human Neuroserpin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7117-U100 21-7117-U500 21-7117-U025 21-7117-U005
10821-002 10820-836 10821-000 10820-998
Recombinant Human Neuroserpin (Carrier-free), Tonbo Biosciences
Neuroserpin, or Serpin I1, is a non-glycosylated inhibitory protein belonging to the serpin superfamily of serine proteinase inhibitors. This brain-associated protein inhibits the tissue-type plasminogen activator (tPA) enzyme. In the central nervous system, tPA is associated with synaptic plasticity and cell death. Neuroserpin expression patterns suggest a role in synaptogenesis. It is secreted by growth cones of neurons and is found predominantly in areas of the brain associated with memory and learning.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty