Human Recombinant Nesfatin 1 (from E. coli)
Supplier: Tonbo Biosciences
21-7114-U100 21-7114-M001 21-7114-U020 21-7114-U500
10820-980EA 801.27 USD
10820-980 10820-830 10820-834 10820-832
Human Recombinant Nesfatin 1 (from E. coli)
Nesfatin-1 is a highly conserved metabolic polypetide derived through cleavage of its precursor protein, nucleobindin 2 (NUCB2). Nesfatin-1 is a secreted protein expressed in the hypothalamus and is implicated in regulation of appetite control and feeding behavior. It is described as an anorexigenic peptide and a satiety molecule. There is some indication that it plays a role in the adaptive response under stress conditions.