Mouse Recombinant Midkine (from E. coli)
Supplier: Tonbo Biosciences
21-7113-U020 21-7113-U005 21-7113-U500 21-7113-U100
10820-828EA 2259.46 USD
10820-828 10820-976 10820-974 10820-978
Mouse Recombinant Midkine (from E. coli)
Midkine, also known as neurite growth-promoting factor 2, is a heparin-binding nonglycosylated low molecular weight protein. It is expressed in a variety of cell types during embryogenesis, and has functions related to cell growth, migration and angiogenesis. Midkine signals through the anaplastic lymphoma kinase (ALK) receptor which stimulates phosphorylation and kinase activation events and results in cell proliferation.