Would you like to log in again?
Recombinant Human sIL-6Rα (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7109-U005 21-7109-U100 21-7109-U500 21-7109-U020
10820-798EA 79.58 USD InStock
10820-798 10820-802 10820-804 10820-800
Recombinant Human sIL-6Rα (Carrier-free), Tonbo Biosciences
Proteins and Peptides
The IL-6 receptor complex is made up of 2 subunits, IL-6R alpha (CD126) and the shared signaling receptor gp130. IL-6 first binds to the low-affinity IL-6R alpha, followed by association with gp130 which then transduces the signal. IL-6R alpha is expressed by many cells including fibroblasts, macrophages and T cells.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty