Would you like to log in again?
Recombinant Human GMF-β (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7108-U002 21-7108-U010 21-7108-U100 21-7108-U500
10820-764EA 86.98 USD InStock
10820-764 10820-766 10820-768 10820-796
Recombinant Human GMF-β (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Glia Maturation Factor beta (GMF-β) is a nerve growth factor and member of the actin-binding proteins ADF family. It is mainly localized in the brain, and functions in the growth and differentiation of neurons and glia. It plays a role in stimulating neural regeneration. GMF-beta has also been found to inhibit the proliferation of a variety of tumor cells in culture.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty