Would you like to log in again?
Recombinant Human Furin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7106-U500 21-7106-U100 21-7106-U010 21-7106-U002
10820-762EA 3159.09 USD InStock
10820-762 10820-760 10820-758 10820-756
Recombinant Human Furin (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Furin is a member of the subtilisin-like proprotein convertase (PC) family, a part of the subtilisin superfamily of serine proteases. It is a type I membrane bound calcium-dependent protease and is expressed in tissues including liver, gut, brain and neuroendocrine. It is localized in the Golgi apparatus and functions to cleave various proproteins with a specific recognition sequence into their mature forms. Substrates are involved in various secretory pathways and include proalbumin, membrane type-1 matrix metalloproteinase, von Willebrand factor and proparathyroid hormone.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty