Would you like to log in again?
Recombinant Human FGFR1a (IIIc) Fc (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7104-U500 21-7104-U050 21-7104-U010 21-7104-U100
10820-746EA 983.02 USD InStock
10820-746 10820-742 10820-740 10820-744
Recombinant Human FGFR1a (IIIc) Fc (Carrier-free), Tonbo Biosciences
Proteins and Peptides
The Fibroblast Growth Factor Receptor (FGFR) family is made up of four known closely related receptor tyrosine kinases, FGFR1 through 4. For FGFR1 through 3, alternative splicing results in numerous variants. One common splicing event results in the extracellular domain having all 3 immunoglobulin-like domains (alpha isoform) or only the II and III Ig domains (beta isoform). Additional variation in the IgIII domain allows for expression as IIIb or IIIc, again due to alternative splicing. Ligand binding results in dimerization and autophosphorylation of specific tyrosine residues. Variants may exhibit different ligand binding properties. FGFR1 predominantly binds the FGF-acidic and FGF-basic proteins.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty