Would you like to log in again?
Recombinant Human EGFR (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7103-U002 21-7103-U500 21-7103-U010 21-7103-U100
10820-732 10820-738 10820-734 10820-736
Recombinant Human EGFR (Carrier-free), Tonbo Biosciences
The EGF Receptor (EGFR) is one of the four members of the EGFR subfamily of receptor tyrosine kinases. It is a transmembrane glycoprotein expressed on the cell surface and is a receptor for EGF and at least six other structurally related ligands including amphiregulin, betacellulin and TGF-alpha. Binding of EGFR to one of these ligands induces receptor dimerization and tyrosine autophosphorylation, leading to cell proliferation. It is involved in regulation of other functions including differentiation, survival, motility and apoptosis. EGFR has been found to be overexpressed in many tumors.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty