Human Recombinant DKK3 (from CHO Cells)
Supplier: Tonbo Biosciences
21-7102-U500 21-7102-U010 21-7102-U100 21-7102-U002
10820-730EA 3789.46 USD
10820-730 10820-724 10820-728 10820-726
Human Recombinant DKK3 (from CHO Cells)
The DKK (Dickkopf) family of secreted proteins includes DKK-1 through -4, and a related protein, Soggy (DKK-11). DKK-3 exhibits the most sequence homology with Soggy. It is expressed in many normal tissues including brain, spinal cord and heart. Also known as REIC (Reduced Expansion in Immortalized Cells), DKK-3 may be a negative growth regulator as expression is downregulated in a variety of cancer cell lines and tumors. Unlike other DKK members, it has been suggested that DKK-3 does not bind to LRP5 and/or LRP6.