Would you like to log in again?
Recombinant Human DKK-1 (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7100-U010 21-7100-U002 21-7100-U100 21-7100-U500
10820-704EA 205.62 USD InStock
10820-704 10820-702 10820-706 10820-714
Recombinant Human DKK-1 (Carrier-free), Tonbo Biosciences
Proteins and Peptides
The DKK (Dickkopf) family of secreted proteins includes DKK-1 through -4, and a related protein, Soggy (DKK-11). DKK-1 is an antagonist of the Wnt signaling pathway, forming inhibitory complexes with Wnt co-receptors LRP5 or LRP6 along with Kremen-1 or Kremen-2. DKK-1 is expressed during development and, in the adult, is expressed in osteoblasts and osteocytes. Over- or under-expression of DKK-1 can result in decreased or increased bone density.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty