Human Recombinant CDNF (from E. coli)
Supplier: Tonbo Biosciences
21-7099-U500 21-7099-U100 21-7099-U025 21-7099-U005
10820-694EA 95.92 USD
10820-694 10820-698 10820-696 10820-700
Human Recombinant CDNF (from E. coli)
Cerebral Dopamine Neurotrophic Factor (CDNF), also known as ARMETL1, is a novel secreted protein that exhibits strong trophic activity on dopaminergic neurons, comparable to that of GDNF. It is structurally similar to MANF. This conserved neurotrophic factor promotes the survival, growth and function of dopaminergic neurons. Besides neurons, it is widely expressed in brain regions including cerebral cortex, cerebellum and hippocampus.