Mouse Recombinant CCL4 (from E. coli)
Supplier: Tonbo Biosciences
21-7098-U500 21-7098-U100 21-7098-U010 21-7098-U002
10820-692EA 2259.6 USD
10820-692 10820-686 10820-688 10820-690
Mouse Recombinant CCL4 (from E. coli)
CCL4 (MIP-1β) is a mitogen-inducible monokine and member of the CC chemokine family. It is a secreted protein that plays a role in the acute inflammatory response and is a chemoattractant for monocytes, neutrophils, eosinophils, NK cells, and CD4+ T lymphocytes. MIP-1β can signal through CCR5 and, by inducing downregulation of this receptor, can block HIV-1 infection of T cells.