Would you like to log in again?
Recombinant Human Artemin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7096-U005 21-7096-U500 21-7096-U100 21-7096-U020
10820-670EA 86.98 USD InStock
10820-670 10820-676 10820-674 10820-672
Recombinant Human Artemin (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Artemin (ARTN) is a secreted disulfide-linked homodimer that belongs to the GDNF family of ligands. It is known to be expressed by embryonic smooth muscle cells and Schwann cells. Artemin promotes sensory neuron survival and likely plays a role in the development of the peripheral nervous system. It signals through the receptor tyrosine kinase (RET)/GFR-alpha 3 receptor complex.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty