Would you like to log in again?
Recombinant Human Amphiregulin (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7095-U050 21-7095-U500 21-7095-U100 21-7095-U010
10820-664EA 194.14 USD InStock
10820-664 10820-668 10820-666 10820-662
Recombinant Human Amphiregulin (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Amphiregulin is a glycoprotein belonging to the EGF family that includes a variety of proteins such as the Neuregulins, TGF-alpha, HB-EGF and Epiregulin. These cytokines are characterized by the presence of at least one EGF structural unit, and all are synthesized as transmembrane precursors which are proteolytically cleaved to produce a soluble form. Amphiregulin stimulates the growth and proliferation of various cell types including keratinocytes, mammary epithelial cells and some fibroblasts, and has also been shown to inhibit growth in certain carcinoma cell lines. Amphiregulin is able to bind to, and signal through, the EGF receptor.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty