Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cart, Core DX, Extended 29" Top, TrippNT

Supplier: TRIPPNT
51807 51421 51296 51807BLUE
10754-746EA 980.56 USD InStock
10754-746 10754-744 10754-612 10754-748
Cart, Core DX, Extended 29" Top, TrippNT
Carts
TrippNT’s 51807 Core DX Lab Cart With Extended 29" Top offers an efficient way to sort, store, organize, lock and transport supplies

  • Core DX Cart with Extended 29 inch top, 6 side shelves, 2 built-in dispensers, 3 extension drawers, and tool holder
  • Durable white polyethylene
  • Drawer: 12 3/4 x 3 5/8 x 15 7/8 WHD
  • Acrylic Sliding Door: comes in 2 colors
  • Delivered fully assembled, no tools required
  • Made in the USA
  • 100% Total Replacement Lifetime Guarantee

It features a larger extended top, 3" non marking nylon rubber casters, 3 full extension drawers and 6 side compartments, 2 with dispensing slots. A large bulk storage area with a locking sliding, clear PETG door secures supplies. 29" x 35" x 19" (width height depth); 66 lb. TrippNT manufactures World Class Carts, Bins and Organizers. With 21 years of experience manufacturing carts, TrippNT is recognized by Inc. 5000 as one of America's fastest growing companies. Polyethylene material. No Security Rail.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty