Would you like to log in again?
FGF-1/FGF-acidic, human recombinant
Supplier: BioVision
4034-10 4034-50 4034-1000
10005-736 10005-740 10005-738
FGF-1/FGF-acidic, human recombinant
An acidic heparin binding growth factor stimulating the proliferation of a variety of cells.
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is <10 ng/ml, corresponding to a specific activity of >1 x 105 units/mg.
FGF-1 is an acidic heparin binding growth factor that stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells. Recombinant human FGF-1 is a 15.8 kDa protein containing 140 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty