Would you like to log in again?
Eotaxin/CCL11, murine recombinant, BioVision
Supplier: BioVision
4029-10 4029-1000
10005-728 10005-730
Eotaxin/CCL11, murine recombinant, BioVision
A chemokine characterized by its high chemotactic selectivity for eosinophils.
The recombinant murine eotaxin was found to induce chemotaxis of purified eosinophils at concentrations ranging between 100 to 1000 ng/ml.
Murine eotaxin shares many of the biological and physical characteristics of human eotaxin. The chemokine exhibits a high chemotactic selectivity for eosinophils and belongs to the C-C family of chemokines, along with chemokines such as RANTES, MCP-3, and MIP-1 α. Murine eotaxin is an 8.4 kDa protein containing 74 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty