Would you like to log in again?
ENA-78, human recombinant, BioVision
Supplier: BioVision
4025-1000 4025-20
10005-724 10005-726
ENA-78, human recombinant, BioVision
A chemokine and activator for neutrophils.
The human ENA-78 produces chemotaxis over a wide concentration range in an assay using human peripheral blood neutrophils. Significant chemotaxis was achieved using a concentration of 50 ng/ml.
ENA-78 (Epithelial Neutrophil Activating Peptide-78) is a recently discovered chemoattractant and activator for neutrophils, belonging to the IL-8 subgroup of the C-X-C family of chemokines. The human ENA-78 is an 8.0 kDa protein containing 74 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty