Would you like to log in again?
EGF, murine recombinant, BioVision
Supplier: BioVision
4023-500 4023-5000 4023-100 4023-1000
10006-018 10005-604 10006-014 10006-016
EGF, murine recombinant, BioVision
A growth factor stimulating cell growth, proliferation, and differentiation by binding to EGFR.
The ED₅₀ as determined by a cell proliferation assay using BALB/c 3T3 cells is < 0.1 ng/ml, corresponding to a specific activity of > 1 x 10⁷ units/mg.
EGF (Epidermal Growth Factor) is polypeptide growth factor that stimulates the proliferation of a wide range of epidermal and epithelial cells. Recombinant murine EGF is a 6.0 kDa protein containing 53 amino acid residues
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty