Would you like to log in again?
Active Sonic Hedgehog (SHH), murine recombinant, BioVision
Supplier: BioVision
4020-100 4020-25 4020-1000
10005-448 10005-452 10005-450
Active Sonic Hedgehog (SHH), murine recombinant, BioVision
Hedgehog signaling pathway ligand involved in regulating vertebrate organogenesis.
Mouse SHH is fully biologically active when compared to standard. Determined by it’s ability to induce alkaline phosphatase production by C3H10T1/2 fibroblasts, with an expected ED₅₀ in the range of 0.03 – 0.30 ug/ml.
Recombinant Mouse Sonic Hedgehog is a member of a small group of secreted proteins that are essential for development in both vertebrates and invertebrates. Three mammalian hedgehog genes (sonic, desert, Indian) share about 60% homology. The Mouse Sonic Hedgehog is 99% homologous to the human gene. Cysteine at position 25 has been substituted with Ile. Mouse SHH is a non-glycosylated polypeptide consisting of 176 amino acids with a molecular weight of 19,800 Da.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty