Would you like to log in again?
CD40Ligand/TRAP, human recombinant, BioVision
Supplier: BioVision
4014-10 4014-50 4014-1000
10005-424 10005-428 10005-426
CD40Ligand/TRAP, human recombinant, BioVision
A protein primarily expressed on activated T cells and involved in immune functions.
Determined by the stimulation of IL-12 induction by human peripheral blood mononuclear cells (PBMC) and the stimulation of IL-8 induction by human PBMC. The expected ED₅₀ for this effect is 5-10 ng/ml. Note: results may vary with PBMC donors.
CD40 Ligand (CD40L/TRAP) is a type II membrane protein that effectuates the helper function of T cells on resting B cells. BioVision’s Human soluble CD40 Ligand is a 16.3 kDa protein containing 149 amino acid residues comprising the receptor binding TNF-like domain of CD40L
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty