Mouse Recombinant C 10 (from E. coli)
Supplier: BioVision
4008-1000 4008-10
10005-408EA 221.82 USD
10005-408 10005-410
Mouse Recombinant C 10 (from E. coli)
A member of the C-C chemokine family related to MIP-1α.
Determined by a chemotaxis assay using mouse spleen MNCs. The maximal activity was achieved at 100 ng/ml.
Murine C-10 is a member of the C-C chemokine family of proteins related to macrophage inflammatory protein-1 α (MIP-1α). The recombinant murine C-10 is a 10.7 kDa protein containing 95 amino acid residues.