Hemorrhoid Forceps, OR Grade, Sklar
Supplier: Sklar
80-1872
10198-804EA 281.42 USD
10198-804
Hemorrhoid Forceps, OR Grade, Sklar
Forceps
Hemorrhoid Forceps are a ratcheted, finger ring instrument used in hemorrhoid procedures, in combination with a hemorrhoidal ligator and rubber bands.
They have curved shanks, that do not obstruct the users view. The serrated tips provide a tight grip on the targeted tissue.