Would you like to log in again?
Baby Mixter Forceps, OR Grade, Sklar
Supplier: Sklar
22-3452C 22-3452 22-3455C 22-3455
75846-942EA 127.67 USD InStock
75846-942 89025-356 75846-944 89025-358
Baby Mixter Forceps, OR Grade, Sklar
Forceps
Baby Mixter Forceps are ratcheted, finger ring forceps used for working in obscured surgical sites
  • Lighter and more delicate
  • Shorter in length
They are most frequently used for clamping, dissection, or grasping tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty