VWR® Performance Series Solid Door Laboratory Refrigerators
SLP-72-TS SLP-36-TS SLP-16-TS SLP-12-TS SLP-26-TS SLP-23-TS SLP-49-TS
10819-332EA 6872.18 USD
10819-332 10820-362 10819-816 10819-338 10819-814 10819-334 10819-336
VWR® Performance Series Solid Door Laboratory Refrigerators
The VWR® Performance Series Solid Door Laboratory Refrigerators feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
  • Data acquisition
  • 24/7 data logging
  • 24-hour digital temp chart
  • Min/max temp history
  • USB port for data transfer
  • 4-20mA output
  • Touchscreen technology
  • Digital temperature display
  • Thermometer bar
  • Electronic owner’s manual access
  • Troubleshooting guide
  • Password protected
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes.
Certifications: UL/C-UL listed.
Ordering information: Unit comes with casters with footed brake. Three-year parts and labor warranty, five years on compressor parts.