Would you like to log in again?
Vernier® Optical DO Probe
Supplier: VERNIER SOFTWARE & TECH SE
ODO-GRD ODO-BTA
470219-314EA 49.99 USD InStock
470219-314 470219-312
Vernier® Optical DO Probe
Data Loggers Sensors
The Vernier Optical DO Probe uses luminescent technology to provide fast, easy, and accurate results, making it a terrific choice for biology, ecology, or environmental science courses. Students can measure dissolved oxygen concentration in surface water or perform a wide variety of experiments to determine changes in dissolved oxygen levels, one of the primary indicators of the quality of an aquatic environment.
Plug-and-play technology; no filling solution, warm-up time, stirring, or calibration necessary
Built-in temperature and pressure compensation
Easy maintenance
Switch setting to measure in units of mg/L or % saturation
The Optical DO Probe cap is warranted to be free from defects for a period of ​two years​ from the date of purchase. With proper use, the lifetime of the cap may be longer than two​ year​s​.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty