DIN to BTA Adapter
DIN-BTA
470219-272EA 5.99 USD
470219-272
DIN to BTA Adapter
This adaptor allows users to connect old DIN style sensors to LabPro, Go!Link, CBL 2, and LabQuest. One end has a female 5-pin DIN connector. The other end is a male BTA connector.