Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Celestron Microscope Imager

Supplier: VERNIER SOFTWARE & TECH SE
CS-DMI
470219-254EA 83.99 USD InStock
470219-254
Celestron Microscope Imager
Microscopes Digital Microscopes
The Celestron Digital Microscope Imager turns traditional compound or stereo microscopes into a high-resolution digital imager using a personal computer, Chromebook, or LabQuest 2. The imager replaces the eyepiece of the microscope and connects directly to the device via USB. With its 2 megapixel sensor, built-in 15x lens, and IR cutoff filter, the Celestron Digital Microscope Imager works together with Logger Pro‘s Image and Video Capture capabilities to provide an easy way to collect still images, time-lapse sequences, and short videos. Chromebook users can use the built-in Camera App in Chrome to take pictures. LabQuest 2 users can use the built-in Camera App in LabQuest App to take pictures as well.

Specifications:
Resolution: 2 megapixel (1600 x 1200 pixels)
Sensor type: CMOS
Eyepiece tube compatibility: 23–30 mm (with included adapter)
Video: 30 fps at VGA resolution (640 x 480)
USB cable length: 2 m

What’s Included:
Celestron Digital Microscope Imager
Adapter collar for use with stereo microscopes
Celestron Digital Microscope Suite software
Instruction manual

Requirements:
Compatible Devices
Windows computers
Windows (XP SP2 and newer)
Celestron Digital Microscope Suite software or Logger Pro software (version 3.8.4 and newer)
Mac computers
Mac (OS X 10.6 and newer)
Celestron Digital Microscope Suite software or Logger Pro software (version 3.8.4 and newer)
Google Chromebooks
Camera app in Chrome
LabQuest 2
Camera App in LabQuest App (version 2.6 and newer)

Not Compatible:
Windows RT devices
iPad, iPhone, or iPod touch
Android devices

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty