Would you like to log in again?
Vernier® Interface Shield
Supplier: VERNIER SOFTWARE & TECH SE
BT-ARD
470219-244EA 30.49 USD InStock
470219-244
Vernier® Interface Shield
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Vernier Arduino Interface Shield plugs directly on top of the Arduino and adds two BTA (analog) and two BTD (digital) sockets. This shield was developed to be used with the SparkFun Arduino RedBoard but will work with the Arduino Uno and other Uno equivalents like the Leonardo. It also works with the Intel Galileo.
Works with over 80 Vernier sensors with BTA and BTD connectors.
Specifications:
Vernier Arduino Interface Shield
2 analog sensor ports (BTA sockets)
2 digital sensor ports (BTD sockets)
Reset button
General-purpose button
Indicator LED
Includes a multiplexer to allow auto-ID to work on both channels (both analog and digital)
Supported Microcontrollers: SparkFun Arduino RedBoard, Arduino UNO, Arduino Leonardo, and Intel Galileo.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty