Would you like to log in again?
Basic Proscope HR Kit, with 50x Lens
Supplier: VERNIER SOFTWARE & TECH SE
BD-HRB
470219-236
Basic Proscope HR Kit, with 50x Lens
The ProScope HR is a USB handheld digital microscope designed for use with a computer. With a high-quality CMOS sensor and universal lens mount, the ProScope HR is a powerful addition to any classroom.
We offer several kits depending on budget and accessories needed. Each kit includes ProScope HR software that allows users to capture still images, time lapse photography, and movies. You can also use the award-winning Logger Pro software (sold separately) to add images to an experiment or lab report to illustrate a concept. Video capture and analysis tools to show live footage and an “instant replay” of an experiment!
Get started on capturing high resolution microscopic images of hair follicles, microwriting on dollar bills, and more with the 50X lens.
ProScope HR base
ProScope HR software
50X len
Image sensor: 1/4″ inch high resolution 1.3 megapixel sensor
Connection type: USB
Image capture resolutions: 320×240 pixels, 640×480 pixels, 1280×1024 pixels
Video capture resolutions: 320×240 pixels @ 30 fps, 640×480 pixels @ 15 fps, 1280×1024 pixels @ 3.75 fps
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty