Reveal G4 Rapid HIV-1 Antibody Test, LAB+, MedMira

Supplier: MedMira
815311007576
10829-304BX 1429.22 USD
10829-304
Reveal G4 Rapid HIV-1 Antibody Test, LAB+, MedMira
Clinical Diagnostic Systems Clinical reagents and kits
Reveal G4 Rapid HIV-1 Antibody Test uses MedMira Rapid Vertical Flow Technology to deliver instant results in three easy steps

Reveal G4 LAB+ features a built-in reagent control line, works with venipuncture whole blood, serum, and plasma specimens, and requires no additional equipment.
Sensitivity: 99.46% Venipuncture Whole Blood, 99.8% Serum, 99.8% Plasma
Specificity: 99.90% Venipuncture Whole Blood, 99.1% Serum, 98.6% Plasma
Test kits can be stored for 12 months at 2 to 30°C (35 to 85°F) and test controls (one positive/one negative) are available as an accessory.

Reveal G4 is the next generation of rapid testing using MedMira Rapid Vertical Flow Technology and research has demonstrated that MedMira's rapid HIV test detects early infection better than other rapid tests and closely matches the performance of much more complicated enzyme immunoassay tests.

HIV-1 Antibody Test Controls are designed for use with Reveal G4 Rapid HIV-1 Antibody Test in customer quality assurance practices. Each set of test controls contains one postive and one negative vial. Each vial is sufficient to perform five Reveal G4 tests. HIV-1 Antibody Test Controls have a shelf life of 12 months, and are stored at room temperature until ready for use.

This simple test provides instant results for either reactive or non-reactive specimens 5 times faster than any lateral flow test. Primary applications for Reveal G4 include occupational exposures, preventing mother-to-child transmission, and emergency departments.

Certifications: FDA Approved.

Ordering information: Each box of Reveal G4 LAB+ includes 50 test kits. Other formats of Reveal G4 are available for testing fingerstick whole blood, serum, and plasma specimens. Test contols are available as an accessory. Each set of HIV-1 Antibody Test Controls contains 1 positive and 1 negative, each sufficient to run five Reveal G4 tests.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR