Molecular Weight Marker, Protein, Pre-stained, Broad Range, 11 to 190 kDa, Thermo Fisher Scientific

J67940-AMI
10816-210EA 215.82 USD
10816-210
Molecular Weight Marker, Protein, Pre-stained, Broad Range, 11 to 190 kDa, Thermo Fisher Scientific
  • Physical Form: Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR