Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercaptobenzothiazole, MALDI Matrix, ultrapure

Supplier: Alfa Aesar
Synonyms: Benzothiazole-2-thiol

J67540-EXK
10816-094PK 111.22 USD InStock
10816-094
2-Mercaptobenzothiazole, MALDI Matrix, ultrapure
2-Mercaptobenzothiazole
For use as a matrix in MALDI mass spectrometry analysis.


Formula: C₇H₅NS₂
MW: 167.26 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 178…182 °C
Density: 1,420
Flash Pt: 243 °C (469 °F)
Storage Temperature: ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty