Chlortetracycline hydrochloride 10 mg/mL in methanol, sterile filtered
|
Danger
|
Synonyms:
7-Chlorotetracycline hydrochloride
, Aureomycin
J67461-AC
J67461-AD
10814-978EA
45.85
USD
10814-978
10814-980
Chlortetracycline hydrochloride 10 mg/mL in methanol, sterile filtered
Chlortetracycline hydrochloride
Formula:
C₂₂H₂₄Cl₂N₂O₈ MW: 515.34 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD26142699 CAS Number: 64-72-2 EINECS: 200-591-7 UN: 1230 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...