Tyrphostin 1 99%

Warning

Synonyms: Tyrphostin A1(4-Methoxybenzylidene)malononitrile2-(4-Methoxybenzylidene)malononitrile((4-Methoxyphenyl)methylene)methane-1,1-dicarbonitrile

J67456-MC J67456-FPL
10814-974EA 607.52 USD
10814-974 10814-972
Tyrphostin 1 99%
Tyrphostin 1
Formula: C₁₁H₈N₂O
MW: 184.198 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00019787
CAS Number: 2826-26-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR