Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

19(R)-Hydroxyprostaglandin E2 0,05% (w/v) in ethanol

Supplier: Alfa Aesar
Danger

J67420-EXG J67420-IAM
10814-966EA 123.28 USD InStock
10814-966 10814-968
19(R)-Hydroxyprostaglandin E2 0,05% (w/v) in ethanol
19(R)-Hydroxyprostaglandin E2


Formula: C₂₀H₃₂O₆
MW: 368.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135220
CAS Number: 64625-54-3
UN: 1170
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty